Learn More
Perkin Elmer US LLC Particle XPRESS Offline Introduction System
Catalog No. 502104111
Encompass_Preferred
Includes: Pump module with 10 mL Syringe 1.5 mm stainless steel sample probe with filtered tip Particle size sensor mount Tubing and completion kit Interface connect cables Power supply with US, UK & European plugs
Catalog No. 50-210-4111 Supplier Perkin Elmer US LLC Supplier No. N0810902
Add to Cart
Add to Cart
Request a Quote
Includes: Pump module with 10 mL Syringe 1.5 mm stainless steel sample probe with filtered tip Particle size sensor mount Tubing and completion kit Interface connect cables Power supply with US, UK & European plugs