Learn More
Perkin Elmer US LLC Parti-Count Particle Count Calibration Fluid, 100 mL
Supplier: Perkin Elmer US LLC N9308731
This Parti-Count Calibration Fluid provides a small aliquot of standard for calibration of the optical sensor and verification of the particle counting system. This Standard contains known amounts of medium test dust at a specified particle size distribution. Volume: Content: 100 mL Calibration Fluid