Learn More
CleanSpace™ Particulate Filter HEPA Hi Capacity
Catalog No. 17800616 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With: PAF-1060, PAF-1070
Catalog No.
17-800-616 For Use With: PAF-1060, PAF-1070
1 options
Catalog No. 17-800-616 Supplier CleanSpace™ Supplier No. PAF1037
Only null left
Add to Cart
Add to Cart
Hi Capacity (HEPA) Particulate Filters are suitable for protection against airborne particulate (dust, mists and fumes).
  • Compatible with ALL CleanSpace™PAPR Power Units
  • CleanSpace™High Efficiency (HEPA) Particulate Filters High Capacity must be used in conjunction with CleanSpace™Filter Adaptor (PAF-0038)
  • Used with the revolutionary CleanSpace™- A light weight PAPR with no hoses/belts
  • Suitable for protection against airborne particulate (dust, mists and fumes).
  • Materials: Fibreglass particulate media and plastic casing, silicone seal
  • Easily fitted and removed from the Power Unit
Specifications
Product Type Filter
For Use With (Equipment) PAF-1060, PAF-1070
Quantity 3/Pk.