Learn More
3M™ N95 Particulate Respirators 8210
SCI_ED
Catalog No. S47062A Shop All 3M Products
Change view
Click to view available options
3m Part Respirator (n95) 20/pk
Catalog No.
S47062A 3m Part Respirator (n95) 20/pk
1 options
This item is not returnable. View return policy
Catalog No. S47062A Supplier 3M™ Supplier No. 7100132742/EMD
This item is not returnable. View return policy
Lightweight disposable particulate respirator designed to provide protection of at least 95 percent filtration efficiency against certain non-oil based particles in applications such as grinding, sanding, sweeping, sawing, bagging or other dusty operations.
  • At least 95% NIOSH tested filter efficiency for use where solids or non-oil-based liquid aerosols are present
  • Synthetic elastic headband
  • Adjustable nose clip provides custom fit and reduces the potential for eyewear fogging
  • Soft nose foam
  • Must be used in accordance with OSHA regulations and the user instructions, warnings and limitations
  • Compatible with a variety of protective eyewear and hearing protection
  • Helps reduce inhalation exposures to certain airborne biological particles (examples: mold, Bacillus anthracis, Mycobacterium tuberculosis)
  • Cannot eliminate the risk of contracting infection, illness or disease 
Order Info
Sold in packs of 20.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0007
Certifications (OSHA) OSHA 1910.134
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Grinding, sanding, sweeping, bagging and other dusty operations
Latex Free Yes
Material Headstrap Thermoplastic Elastomer
Model 8210/EMD
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Quantity 160/Cs.
Sterility Non-sterile
With Faceseal No
Strap Attachment Type Two-Strap Design
FDA Cleared Surgical Mask No