Learn More
SCP Science PartiStan 2806 Calibration Fluid, 400 mL
Catalog No. NC9040399
Encompass
Designed for secondary calibrations of automatic particle counters (APCs) as per ISO 11171. Partistan 2806 Calibration Fluid is a secondary calibration fluid traceable to NIST SRM 2806b.
Catalog No. NC9040399 Supplier SCP Science Supplier No. 150701001
Designed for secondary calibrations of automatic particle counters (APCs) as per ISO 11171. Partistan 2806 Calibration Fluid is a secondary calibration fluid traceable to NIST SRM 2806b.