Learn More
SCP Science CONOSTAN PartiStan 2806 Calibration Fluid, Secondary Calibration, ISO 11171 Compliance, 400 mL
Catalog No. NC1209768
Encompass
PartiStan 2806 Calibration Fluid (400 mL) is a specialized calibration fluid from CONOSTAN, designed for precise secondary calibration of automatic particle counters (APCs). It complies with ISO 11171 standards and ensures NIST traceability, offering reliable performance for specific applications where reduced concentration is required. Ideal for laboratories, it supports accurate measurement and enhances compliance with industry standards, setting it apart from standard calibration fluids.
Catalog No. NC1209768 Supplier SCP Science Supplier No. 150701008
PartiStan 2806 Calibration Fluid (400 mL) is a specialized calibration fluid from CONOSTAN, designed for precise secondary calibration of automatic particle counters (APCs). It complies with ISO 11171 standards and ensures NIST traceability, offering reliable performance for specific applications where reduced concentration is required. Ideal for laboratories, it supports accurate measurement and enhances compliance with industry standards, setting it apart from standard calibration fluids.