Learn More
Graphic Controls LLC Partlow 7 Day Circular Chart - 100 per box
Catalog No. NC3523396
Encompass_Preferred
Partlow 7 Day Circular Chart - 100 per box Chart Size 09.938" Time Rotation 7D Ranges 0 - 100 Uniform along the Arc (0 TO 100 BY 10 ) To Fit Recorder Models: MRC 7000
Catalog No. NC3523396 Supplier Graphic Controls LLC Supplier No. 1115328
Partlow 7 Day Circular Chart - 100 per box Chart Size 09.938" Time Rotation 7D Ranges 0 - 100 Uniform along the Arc (0 TO 100 BY 10 ) To Fit Recorder Models: MRC 7000