Learn More
Grainger SHUTER Parts Cabinet, Plastic, 20 Bin, 1 ea
Catalog No. 502774951
Encompass_Preferred
This small parts drawer cabinet features a durable plastic frame and drawers that resist rust. Designed for efficient organization, the unit includes 20 bins and is suitable for benchtop use. Cabinets can be stacked with compatible models of the same width and depth to create a high-density storage system.
  • Features 20 integrated storage bins
  • Constructed from corrosion-resistant plastic
  • Designed for stackable, high-density storage configurations
  • Dimensions: 14-3/4 in. wide by 7-3/8 in. high by 6 in. deep
  • Bench style cabinet suitable for workspaces
Catalog No. 50-277-4951 Supplier Grainger Supplier No. 172J59
This small parts drawer cabinet features a durable plastic frame and drawers that resist rust. Designed for efficient organization, the unit includes 20 bins and is suitable for benchtop use. Cabinets can be stacked with compatible models of the same width and depth to create a high-density storage system.
  • Features 20 integrated storage bins
  • Constructed from corrosion-resistant plastic
  • Designed for stackable, high-density storage configurations
  • Dimensions: 14-3/4 in. wide by 7-3/8 in. high by 6 in. deep
  • Bench style cabinet suitable for workspaces