Learn More
Mac-Mod MarvelXACT SS 100µm x 150mm supplied by MAC-MOD
SDP
Catalog No. 5000152311
Encompass
MAC-MOD helps you ensure a perfect connection every time with the MarvelXACT™ Fitting System - the options available are:No risk of under- or over-tightening with torque limiting mechanismFace sealing ensures Zero Dead VolumeMinimize carryover, peak broadening, and peak tailingFinger-tight to 19,000 psi100+ connects and disconnectsNo training or instructions needed to make a perfect connection everytimeBioinert BioVersal™ PEEK Lined Stainless Steel and PEEKsil Configurations Available
Catalog No. 50-001-52311 Supplier Mac-Mod Supplier No. MMUPFS7100150
MAC-MOD helps you ensure a perfect connection every time with the MarvelXACT™ Fitting System - the options available are:No risk of under- or over-tightening with torque limiting mechanismFace sealing ensures Zero Dead VolumeMinimize carryover, peak broadening, and peak tailingFinger-tight to 19,000 psi100+ connects and disconnectsNo training or instructions needed to make a perfect connection everytimeBioinert BioVersal™ PEEK Lined Stainless Steel and PEEKsil Configurations Available