Learn More
Microsolv Technology Corporation Partsil 10Um Scx 250X4.6Mm 1Ea
Supplier: Microsolv Technology Corporation 6318625PW
Hichrom Partisil 10µm SCX column, 250 x 4.6mm (WCS hardware), HPLC Column, (formerly Whatman GE). 1 each.