Learn More
Aviva Systems Biology Corp PARVB Antibody - C-terminal region (ARP46018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARVB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PARVB. Alternate names: CGI-56ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59179