Learn More
Drg International Inc DRG Parvovirus B19 IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3503R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Parvovirus B19 in serum. This product is for Research Use Only. The DRG Parvovirus B19 IgG (Recombinant) ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with recombinant Parvovirus B19 antigen (VP1 proteins). Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7581