Learn More
Drg International Inc DRG Parvovirus B19 IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3504R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Parvovirus B19 in serum. This product is for Research Use Only. The DRG Parvovirus B19 IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with Parvovirus B19 antigen. Pretreated specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7582