Learn More
IMEB, Inc PAS-Periodic/Schiff Stain Kit. 8 Oz Bottle/Solutions of: Periodic Acid, Schiff’s Reagent, Gold Chloride, Hematoxlyin Nuclear Stain, Light Green SF Counterstain, 5-2 Oz. Staining Containers and Tray.
SDP
Catalog No. 501689474
Encompass_Preferred
Periodic/Schiff’s Stain Kit is used to stain Fungus, Mucin Glycogen , and Basement Membranes. The result is bright pink staining with a green background and black to deep purple nuclei. Contains: 1- 8 oz. Bottle Periodic Acid solution, 1- 8 oz. Bottle Schiff’s Reagent solution, 1- 8 oz. Bottle Gold Chloride solution, .1- 8 oz. Bottle Hematoxlyin Nuclear stain solution, 1- 8 oz. Bottle Light Green SF counterstain, 5- 2 oz. Staining containers, and storage tray.
Catalog No. 50-168-9474 Supplier IMEB, Inc Supplier No. K7308
Periodic/Schiff’s Stain Kit is used to stain Fungus, Mucin Glycogen , and Basement Membranes. The result is bright pink staining with a green background and black to deep purple nuclei. Contains: 1- 8 oz. Bottle Periodic Acid solution, 1- 8 oz. Bottle Schiff’s Reagent solution, 1- 8 oz. Bottle Gold Chloride solution, .1- 8 oz. Bottle Hematoxlyin Nuclear stain solution, 1- 8 oz. Bottle Light Green SF counterstain, 5- 2 oz. Staining containers, and storage tray.