Learn More
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit, 500 ml kit
SDP
Catalog No. NC9990422
Encompass
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit procedure, with methods for glycogen digestion, is used for staining glycoproteins and may aid in the differential diagnosis of tumors through detection of acid/neutral epithelial mucins and/or glycogen.   Digestion steps can be employed for further identification of mucosubstances.  PAS can also be used for staining basement membranes and fungal cell walls.
Catalog No. NC9990422 Supplier Newcomer Supply Supplier No. 9162B
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit procedure, with methods for glycogen digestion, is used for staining glycoproteins and may aid in the differential diagnosis of tumors through detection of acid/neutral epithelial mucins and/or glycogen.   Digestion steps can be employed for further identification of mucosubstances.  PAS can also be used for staining basement membranes and fungal cell walls.