Learn More
StatLab PAS With and Without Digestion Control Slides
SDP
Catalog No. 21033345
Change view
Click to view available options
Form:
Solid
Catalog No. Form
21-033-345 Solid
1 options
Catalog No. 21-033-345 Supplier StatLab Supplier No. CSPAS/25
Add to Cart
Add to Cart
Use PAS control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Form Solid
Product Type Periodic Acid Schiff