Learn More
GSC Go Science Crazy Pascal's Ball
SCI_ED
Catalog No. S28119 Shop All GSC Products
Encompass
Click to view available options
:
Pascal's ball
Not a toy. Product intended for ages 13 and up.
Not a toy. Product intended for ages 13 and up.
Demonstration to help students understand Pascal's Law.
Pascal's Ball is filled with water and then force is applied to the plunger. The water will then follow Pascal's Law and will exit the ball with equal force in all directions.
  • Visually demonstrate Pascal's Law
  • Includes instructions
Specifications
Product Type Pascal's Ball
Includes instructions
For Use With (Application) Helps students understand Pascal' law
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.