Learn More
Apexbio Technology LLC Pasiniazid 2066-89-9 1g
SDP
Catalog No. 5000095246
Encompass_Preferred
Pasiniazid (CAS 2066-89-9) is a small-molecule inhibitor targeting enzymes essential for mycolic acid biosynthesis in mycobacteria It is designed to disrupt cell wall synthesis thereby inhibiting the growth of Mycobacterium tuberculosis and Mycobacterium leprae Pasiniazid exerts its biological activity primarily through inhibition of mycolic acid biosynthesis In in vitro antimicrobial assays and animal infection models Pasiniazid demonstrates inhibitory activity with reported IC50 values against mycobacterial targets typically within micromolar concentrations Based on these pharmacological properties Pasiniazid holds research potential in the investigation of tuberculosis and leprosy pathogenesis including studies on drug-response relationships resistance mechanisms and pharmacokinetics
Catalog No. 50-000-95246 Supplier Apexbio Technology LLC Supplier No. A84971000
Pasiniazid (CAS 2066-89-9) is a small-molecule inhibitor targeting enzymes essential for mycolic acid biosynthesis in mycobacteria It is designed to disrupt cell wall synthesis thereby inhibiting the growth of Mycobacterium tuberculosis and Mycobacterium leprae Pasiniazid exerts its biological activity primarily through inhibition of mycolic acid biosynthesis In in vitro antimicrobial assays and animal infection models Pasiniazid demonstrates inhibitory activity with reported IC50 values against mycobacterial targets typically within micromolar concentrations Based on these pharmacological properties Pasiniazid holds research potential in the investigation of tuberculosis and leprosy pathogenesis including studies on drug-response relationships resistance mechanisms and pharmacokinetics