Learn More
Apexbio Technology LLC Pasireotide Acetate 396091-76-2 5mg
SDP
Catalog No. 5000090741
Encompass_Preferred
Pasireotide Acetate (CAS 396091-76-2) is a cyclohexapeptide somatostatin analog that acts as a multi-receptor ligand selectively activating several somatostatin receptor subtypes (sst1/2/3/5 with pKi values of 8 2 9 0 9 1 and 9 9 respectively) By modulating these receptors Pasireotide Acetate regulates the secretion of growth hormone insulin-like growth factor I (IGF-I) and adrenocorticotropic hormone (ACTH) at the cellular level Preclinical investigations indicate its capacity to alter endocrine hormone release with implications for studies in acromegaly and Cushing s disease Its demonstrated effects on secretion inhibition proliferation control and induction of apoptosis make it a valuable tool in studies of related biological pathways
Catalog No. 50-000-90741 Supplier Apexbio Technology LLC Supplier No. B35965
Pasireotide Acetate (CAS 396091-76-2) is a cyclohexapeptide somatostatin analog that acts as a multi-receptor ligand selectively activating several somatostatin receptor subtypes (sst1/2/3/5 with pKi values of 8 2 9 0 9 1 and 9 9 respectively) By modulating these receptors Pasireotide Acetate regulates the secretion of growth hormone insulin-like growth factor I (IGF-I) and adrenocorticotropic hormone (ACTH) at the cellular level Preclinical investigations indicate its capacity to alter endocrine hormone release with implications for studies in acromegaly and Cushing s disease Its demonstrated effects on secretion inhibition proliferation control and induction of apoptosis make it a valuable tool in studies of related biological pathways