Learn More
Medchemexpress LLC Pasireotide acetate | 396091-76-2 | 99.5% | 1107.26 | 1 MG
SDP
Catalog No. 5000393829
Encompass_Preferred
Pasireotide acetate is a long-acting cyclohexapeptide somatostatin analogue. It enhances agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5). This compound can suppress Growth Hormone (GH), Insulin-like Growth Factor-I (IGF-I), and Adrenocorticotropic Hormone (ACTH) secretion, indicating potential effectiveness in treating acromegaly and Cushing's disease.
  • Enhances agonist activity at somatostatin receptors
  • Suppresses GH, IGF-I, and ACTH secretion
  • Exhibits antisecretory, antiproliferative, and proapoptotic activities
  • Effectively inhibits GHRH-induced GH release in primary rat pituitary cells
  • Decreases serum insulin and increases serum glucose in vivo
  • Reduces tumor size and increases apoptosis in Pdx1-Cre mice
  • Exerts antinociceptive and antiinflammatory actions via SSTR2 receptor
Catalog No. 50-003-93829 Supplier Medchemexpress LLC Supplier No. HY16381A1MG
Pasireotide acetate is a long-acting cyclohexapeptide somatostatin analogue. It enhances agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5). This compound can suppress Growth Hormone (GH), Insulin-like Growth Factor-I (IGF-I), and Adrenocorticotropic Hormone (ACTH) secretion, indicating potential effectiveness in treating acromegaly and Cushing's disease.
  • Enhances agonist activity at somatostatin receptors
  • Suppresses GH, IGF-I, and ACTH secretion
  • Exhibits antisecretory, antiproliferative, and proapoptotic activities
  • Effectively inhibits GHRH-induced GH release in primary rat pituitary cells
  • Decreases serum insulin and increases serum glucose in vivo
  • Reduces tumor size and increases apoptosis in Pdx1-Cre mice
  • Exerts antinociceptive and antiinflammatory actions via SSTR2 receptor