Learn More
Medchemexpress LLC Pasireotide acetate | 396091-76-2 | 99.5% | 1107.26 | 25 MG
SDP
Catalog No. 5000395439
Encompass_Preferred
Pasireotide (SOM230) acetate is a long-acting cyclohexapeptide somatostatin analogue that enhances agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5). It can suppress growth hormone, IGF-I, and ACTH secretion, indicating potential efficacy in treating acromegaly and Cushing's disease. This compound also demonstrates antisecretory, antiproliferative, and proapoptotic activities.
  • Exhibits unique high-affinity binding to human somatostatin receptors.
  • Effectively inhibits growth hormone releasing hormone (GHRH) induced growth hormone (GH) release in primary cultures of rat pituitary cells.
  • Significantly decreases serum insulin and increases serum glucose.
  • Reduces tumor size and increases apoptosis.
  • Exerts antinociceptive and antiinflammatory actions via the SSTR2 receptor.
Catalog No. 50-003-95439 Supplier Medchemexpress LLC Supplier No. 25MGHY16381A
Pasireotide (SOM230) acetate is a long-acting cyclohexapeptide somatostatin analogue that enhances agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5). It can suppress growth hormone, IGF-I, and ACTH secretion, indicating potential efficacy in treating acromegaly and Cushing's disease. This compound also demonstrates antisecretory, antiproliferative, and proapoptotic activities.
  • Exhibits unique high-affinity binding to human somatostatin receptors.
  • Effectively inhibits growth hormone releasing hormone (GHRH) induced growth hormone (GH) release in primary cultures of rat pituitary cells.
  • Significantly decreases serum insulin and increases serum glucose.
  • Reduces tumor size and increases apoptosis.
  • Exerts antinociceptive and antiinflammatory actions via the SSTR2 receptor.