Learn More
Electron Microscopy Sciences AMP Passive Air Isolation Platform with White Epoxy Surface, 90 x 90 cm
SDP
Catalog No. 5034447
Encompass_Preferred
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 90 x 90 cm
Surface: White Epoxy
Catalog No. 50-344-47 Supplier Electron Microscopy Sciences Supplier No. 6390
These High Performance anti-vibration platforms for microscopes are designed to be pumped-up and left alone. Simple to use, they are ideal for use with fixed loads or in applications where pressurised air or gas cannot be used. The unique air damped rolling diaphragm sub 2Hz frequency isolator removes up to 99 percent of vibrations. And sound deadening construction eliminates unwanted surface vibrations. They support loads up to 100kg .
Size: 90 x 90 cm
Surface: White Epoxy