Learn More
Carnabio Usa Inc PASK/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0212810
Encompass
PASK, Catalytic domain, Wild type, Amino Acid 949-1323, NP_055963.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0920