Learn More
Eurofins Discoverx PASK, active, 250 µg. (14-701M)
Supplier:  Eurofins Discoverx 14701M
Encompass
6xHis-tagged recombinant, human full length PASK amino acids 995–end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9214