Learn More
Omnimed Inc Pass-Thru Narcotic Cabinets (5"- 7.25") Wall Thickness
SDP
Supplier:  Omnimed Inc 181786
Encompass_Preferred
The Omnimed Pass-Thru Stainless Steel Cabinet offers a durable, secure solution for transferring specimens between patient areas and the laboratory. Constructed from heavy-duty stainless steel, this cabinet is designed for long-lasting use and provides a reliable means of transporting medical samples. The adjustable trim flange allows the cabinet to fit snugly into walls with thicknesses ranging from 5" to 7.25", securing it securely in place. Both sides feature self-closing doors with a robust piano hinge spring closure, and the unit is equipped with key lock security to protect specimens. The locks are keyed alike and come with three keys for easy access. Additionally, the ambidextrous top lifts to allow for flexible left or right door access, making the unit adaptable to different setups. Backed by a 10-year warranty (1-year on locks), this pass-thru cabinet ensures reliable performance for years to come.
Catalog No. NC3956389