Learn More
Mitegen Passive Drying and Storage Rack for 4L LN2 Dewars
SDP
Catalog No. 5000495011
Encompass
Keep your lab organized and your 4L liquid nitrogen (LN2) dewars secure with the convenient Passive Drying and Storage Rack. This compact and stable stand provides a home for your dewars after cleaning procedures. This rack is the perfect companion to the Active 4L Dewar Drying Rack SKU M-CEM-NS-ADR-LD4-1, which uses heated, filtered air to completely dry the inside of your dewar in just 35 minutes. By pairing the two products, you can create a streamlined and efficient workflow for your lab's cryogenic storage. The rack offers an easy way to store your 4L dewars, ensuring they are always ready for use.
Catalog No. 50-004-95011 Supplier Mitegen Supplier No. MCEMNSPDRLD41
Keep your lab organized and your 4L liquid nitrogen (LN2) dewars secure with the convenient Passive Drying and Storage Rack. This compact and stable stand provides a home for your dewars after cleaning procedures. This rack is the perfect companion to the Active 4L Dewar Drying Rack SKU M-CEM-NS-ADR-LD4-1, which uses heated, filtered air to completely dry the inside of your dewar in just 35 minutes. By pairing the two products, you can create a streamlined and efficient workflow for your lab's cryogenic storage. The rack offers an easy way to store your 4L dewars, ensuring they are always ready for use.