Learn More
Waters Corp Passive PreHeater, MP35N, 0.005 in. ID
Catalog No. 502392844 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement Passive PreHeater (PPH) designed for use in Arc Bio 3cm Column Heater or Heater/Coolers. It is constructed from MP35N material and features a 0.005 in. internal diameter.
  • Constructed from MP35N material
  • Features a 0.005 in. internal diameter
  • Designed for use in Arc Bio 3cm column heater or heater/coolers
Catalog No. 50-239-2844 Supplier Waters Corp Supplier No. 700011699
This is a replacement Passive PreHeater (PPH) designed for use in Arc Bio 3cm Column Heater or Heater/Coolers. It is constructed from MP35N material and features a 0.005 in. internal diameter.
  • Constructed from MP35N material
  • Features a 0.005 in. internal diameter
  • Designed for use in Arc Bio 3cm column heater or heater/coolers