Learn More
Agilent Technologies PASSIVE REFERENCE DYE
Catalog No. 50125990
Encompass
Passive Reference Dye, QPCR Reagents, Reagent for any fluorescent chemistry, Wide linear dynamic range of amplification, Passive reference dye included in a separate tube to maximize performance on different instrument platforms
Catalog No. 50-125-990 Supplier Agilent Technologies Supplier No. 600536
Passive Reference Dye, QPCR Reagents, Reagent for any fluorescent chemistry, Wide linear dynamic range of amplification, Passive reference dye included in a separate tube to maximize performance on different instrument platforms