Learn More
Euthanex Vacuum Interface for Passive Exhausting of Halogenated Anesthetics.
Catalog No. 501540178
Encompass_Preferred
Vacuum Interface. Connects mouse and rat breathers or induction chamber to house vacuum system. Allows these devices to exhaust halogenated anesthetic gases passively without interfering with the anesthesia procedure. Can accommodate waste gas from up to two devices. Choose from four interchangeable exhaust tube connections: 1/8", 1/4", 1/2" or 22mm inner diameter.
Catalog No. 50-154-0178 Supplier Euthanex Supplier No. EZ240
Vacuum Interface. Connects mouse and rat breathers or induction chamber to house vacuum system. Allows these devices to exhaust halogenated anesthetic gases passively without interfering with the anesthesia procedure. Can accommodate waste gas from up to two devices. Choose from four interchangeable exhaust tube connections: 1/8", 1/4", 1/2" or 22mm inner diameter.