Learn More
SPS Medical PASSPORT MAIN-IN SYSTEM 12/EA
Catalog No. NC9573110
Encompass_Preferred
PassPort System, 12 tests/bx , Description:A mail-in, sterilizer BI monitoring service designed to provide 3rd party, independent verification that sterilization was achieved , Function:For use in Steam
Catalog No. NC9573110 Supplier SPS Medical Supplier No. PS012
PassPort System, 12 tests/bx , Description:A mail-in, sterilizer BI monitoring service designed to provide 3rd party, independent verification that sterilization was achieved , Function:For use in Steam