Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 51 x 32 x 11 mm
SDP
Catalog No. 5027719
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 51 x 32 x 11 mm
Dimensions: 53 x 37 x 15 mm
Catalog No. 50-277-19 Supplier Electron Microscopy Sciences Supplier No. 6432602
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 51 x 32 x 11 mm
Dimensions: 53 x 37 x 15 mm