Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 63 x38 x 15 mm
SDP
Catalog No. 5027717
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 63 x 38 x 15 mm
Dimensions: 66 x 44 x 18 mm
Catalog No. 50-277-17 Supplier Electron Microscopy Sciences Supplier No. 6432604
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 63 x 38 x 15 mm
Dimensions: 66 x 44 x 18 mm