Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 76 x 43 x 18 mm
SDP
Catalog No. 5027716
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 76 x 43 x 18 mm
Dimensions: 78 x 48 x 20 mm
Catalog No. 50-277-16 Supplier Electron Microscopy Sciences Supplier No. 6432606
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 76 x 43 x 18 mm
Dimensions: 78 x 48 x 20 mm