Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 80 x 53 x 31 mm
SDP
Catalog No. 5027715
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 80 x 53 x 31 mm
Dimensions: 82 x 57 x 34 mm
Catalog No. 50-277-15 Supplier Electron Microscopy Sciences Supplier No. 6432611
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 80 x 53 x 31 mm
Dimensions: 82 x 57 x 34 mm