Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 82 x 51 x 22 mm
SDP
Catalog No. 5027721
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 82 x 51 x 22 mm
Dimensions: 85 x 54 x 25 mm
Catalog No. 50-277-21 Supplier Electron Microscopy Sciences Supplier No. 6432612
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 82 x 51 x 22 mm
Dimensions: 85 x 54 x 25 mm