Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 65 x 40 x 25 mm
SDP
Catalog No. 5027722
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 65 x 40 x 25 mm
Dimensions: 70 x 45 x 28 mm
Catalog No. 50-277-22 Supplier Electron Microscopy Sciences Supplier No. 6432605
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 65 x 40 x 25 mm
Dimensions: 70 x 45 x 28 mm