Learn More
Electron Microscopy Sciences Sliding Paste Board Box, 72 x 47 x 28 mm
SDP
Catalog No. 5027712
Encompass_Preferred
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 72 x 47 x 28 mm
Dimensions: 75 x 51 x 30 mm
Catalog No. 50-277-12 Supplier Electron Microscopy Sciences Supplier No. 6432610
Constructed from white pasteboard; match box style and rectangular in shape. It is ideal for the storage of specimen blocks and many small items.
Interior: 72 x 47 x 28 mm
Dimensions: 75 x 51 x 30 mm