Learn More
Med Vet International Coloplast Triad™ Hydrophilic Wound Dressing Paste, 6oz, 1967, Each
Catalog No. 503043688
Encompass_Preferred
Hydrophilic Wound Dressing Paste Triad™ 6 oz. Zinc oxide-based hydrophilic paste gently adheres to moist wound beds Autolytic action loosens dried eschar to aid in its removal process Sterile, Non-adhesive dressing gently covers moist, weeping wounds and spreads evenly over entire wound area
Catalog No. 50-304-3688 Supplier Med Vet International Supplier No. 227283EA
Add to Cart
Add to Cart
Hydrophilic Wound Dressing Paste Triad™ 6 oz. Zinc oxide-based hydrophilic paste gently adheres to moist wound beds Autolytic action loosens dried eschar to aid in its removal process Sterile, Non-adhesive dressing gently covers moist, weeping wounds and spreads evenly over entire wound area