Learn More
Bellco Glass, Inc. Pasteur Pipet,5x3"
SDP
Catalog No. 501944646
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Bent tip with 1.5 to 2mm orifice. MADE IN THE USA Borosilicate Glass. Case qty-144
Catalog No. 50-194-4646 Supplier Bellco Glass, Inc. Supplier No. 127350003
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Bent tip with 1.5 to 2mm orifice. MADE IN THE USA Borosilicate Glass. Case qty-144