Learn More
Bellco Glass, Inc. Pasteur Pipet,5X3"
SDP
Catalog No. 501944643
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Straight tip with 1.5 to 2mm orifice. Borosilicate Glass. MADE IN THE USA Case qty-288
Catalog No. 50-194-4643 Supplier Bellco Glass, Inc. Supplier No. 127150003
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Straight tip with 1.5 to 2mm orifice. Borosilicate Glass. MADE IN THE USA Case qty-288