Learn More
Bellco Glass, Inc. Pasteur Pipet, 6X2" 8mm OD"
SDP
Catalog No. 501944644
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Straight tip with 1.5 to 2mm orifice. Borosilicate Glass. MADE IN THE USA Case qty-288
Catalog No. 50-194-4644 Supplier Bellco Glass, Inc. Supplier No. 127160002
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.Straight tip with 1.5 to 2mm orifice. Borosilicate Glass. MADE IN THE USA Case qty-288