Learn More
Research Products International Corp Disposable Plastic Pasteur Pipets, 5 3/4 Inch, Sterile, Individually Wrapped, 50 per Bag, 200 per Case
SDP
Catalog No. 501367940
Encompass
Disposable plastic pasteur pipets offer the same aspirating precision of glass pipets with the added safety benefits of plastic. Plastic construction provides a stem that withstands flexing without the risk of breakage. Pipets can be used with aspirating bulbs or attached to a vacuum apparatus. The pipets are individually wrapped and packaged 50 to a bag with 200 pipets per case.
Catalog No. 50-136-7940 Supplier Research Products International Corp Supplier No. 248607
Disposable plastic pasteur pipets offer the same aspirating precision of glass pipets with the added safety benefits of plastic. Plastic construction provides a stem that withstands flexing without the risk of breakage. Pipets can be used with aspirating bulbs or attached to a vacuum apparatus. The pipets are individually wrapped and packaged 50 to a bag with 200 pipets per case.