Learn More
Sigma Aldrich Fine Chemicals Biosciences Pasteur pipettes short cap
Catalog No. 502900785
Encompass
Soda-lime-glass (ISO 7712). Long drawn-out capillary tip. Suction end with constriction for a cotton plug.
Catalog No. 50-290-0785 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6279921000EA
Add to Cart
Add to Cart
Soda-lime-glass (ISO 7712). Long drawn-out capillary tip. Suction end with constriction for a cotton plug.