Learn More
Braintree Scientific Pasteur Transfer Pipette,1 ml, 104mm, 17-22ul, non-sterile
SDP
Catalog No. 501955521
Encompass
Transfer Pipettes - Single-use Pasteur pipettes. Our range includes a large variety of transfer pipettes for safe and convenient application when transferring liquids in laboratories. Disposable Pasteur pipettes are suited for versatile application without any risk of injury or infection from broken glass. As single-use products, pipettes safely prevent carry over or cross-contamination. Practical graduations are ideally suited for sample measurement and transfer. Pipettes can be heat-sealed to store residual volumes. For a sterile transfer of liquids, our range includes sterile options either individually wrapped or 20 pipettes per pack. Polyethylene material is environmentally neutral thereby ensuring non-toxic disposal. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5521 Supplier Braintree Scientific Supplier No. PIP1180
Transfer Pipettes - Single-use Pasteur pipettes. Our range includes a large variety of transfer pipettes for safe and convenient application when transferring liquids in laboratories. Disposable Pasteur pipettes are suited for versatile application without any risk of injury or infection from broken glass. As single-use products, pipettes safely prevent carry over or cross-contamination. Practical graduations are ideally suited for sample measurement and transfer. Pipettes can be heat-sealed to store residual volumes. For a sterile transfer of liquids, our range includes sterile options either individually wrapped or 20 pipettes per pack. Polyethylene material is environmentally neutral thereby ensuring non-toxic disposal. **Sold for research purposes only and not for human use**