Learn More
Innovative Research Inc Pooled Human Pasteurized Breast Milk 50 ml
Supplier:  Innovative Research Inc IRHUBMKPST50ML
Encompass_Preferred
Pooled HumanPasteurized Breast Milk from Innovative Research is pateurized at 72°C for 15 seconds. It is pooled with a minimum of three donors.
Catalog No. 50-203-6043