Learn More
Chemglass Life Sciences Pasteur Pipette, 3mL, Individually Wrapped, Sterile
SDP
Catalog No. 501897695
Encompass_Preferred
Material: Medical Grade Transparent LDPE. Excellent transparency with clear graduations for easy observation. Good fluidity for easy control. High precision and drop repeatability. Individually wrapped. Sterilized by E-beam. DNase/RNase, endotoxin and pyrogenic-free.
Catalog No. 50-189-7695 Supplier Chemglass Life Sciences Supplier No. CGN3404103
Material: Medical Grade Transparent LDPE. Excellent transparency with clear graduations for easy observation. Good fluidity for easy control. High precision and drop repeatability. Individually wrapped. Sterilized by E-beam. DNase/RNase, endotoxin and pyrogenic-free.