Learn More
Organomation One Dozen Pasteur Pipet Adapters with 1/8 Hose Barb Fitting
SDP
Catalog No. NC0602332
Encompass_Preferred
These adapters allow Pasteur pipettes to be used instead of stainless steel needles. This is the male luer to 1/8 hose barb model compatible with all Organomation nitrogen evaporators. Set of one dozen.
Catalog No. NC0602332 Supplier Organomation Supplier No. NA0637
These adapters allow Pasteur pipettes to be used instead of stainless steel needles. This is the male luer to 1/8 hose barb model compatible with all Organomation nitrogen evaporators. Set of one dozen.