Learn More
TARGETMOL CHEMICALS INC PAT-1251 5MG
SDP
Catalog No. 502581462
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PAT-1251 is an inhibitor of LOXL2 with IC50s of 10 0.12 and 0.16 uM for the mouse rat and dog and can be used in studies about fibrotic diseases. purity: 99%
Catalog No. 50-258-1462 Supplier TARGETMOL CHEMICALS INC Supplier No. T12371L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PAT-1251 is an inhibitor of LOXL2 with IC50s of 10 0.12 and 0.16 uM for the mouse rat and dog and can be used in studies about fibrotic diseases. purity: 99%