Learn More
Medchemexpress LLC HY-107781 5mg Medchemexpress, PAT-505 CAS:1782070-22-7 Purity:>98%
SDP
Catalog No. 501932297
Encompass_Preferred
Medchemexpress, HY-107781 5mg PAT-505 CAS:1782070-22-7 PAT-505 is a potent, selective, noncompetitive and orally available autotaxin inhibitor, with an IC 50 of 2 nM in Hep3B cells, 9.7 nM in human blood and 62 nM in mouse plasma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2297 Supplier Medchemexpress LLC Supplier No. HY1077815MG
Medchemexpress, HY-107781 5mg PAT-505 CAS:1782070-22-7 PAT-505 is a potent, selective, noncompetitive and orally available autotaxin inhibitor, with an IC 50 of 2 nM in Hep3B cells, 9.7 nM in human blood and 62 nM in mouse plasma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.