Learn More
ERA WATERS CORP PAT700 Calibration Standards Kit
SDP
Catalog No. NC1680973
Encompass
Set of four bottles includes (1) Blank, (1) 0.250 mg/L C NIST Sucrose, (1) 0.500 mg/L C NIST Sucrose, (1) 0.750 mg/L C NIST Sucrose in 60 mL bottles with RFID Tags.
Catalog No. NC1680973 Supplier ERA WATERS CORP Supplier No. 19202
Add to Cart
Edge
Add to Cart
Set of four bottles includes (1) Blank, (1) 0.250 mg/L C NIST Sucrose, (1) 0.500 mg/L C NIST Sucrose, (1) 0.750 mg/L C NIST Sucrose in 60 mL bottles with RFID Tags.